Anti-IgG Rat Monoclonal Antibody [clone: LO-MG2a-7] (Biotin)

Supplier: GeneTex
GTX11590
89307-122EA 374.6 USD
89307-122
Anti-IgG Rat Monoclonal Antibody [clone: LO-MG2a-7] (Biotin)
Antibodies
Recommended Starting Dilutions:ELISA: Use at a concentration of 0.5 - 1 µg/mL.Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: LO-MG2a-7
Conjugation: Biotin
Epitope: Heavy Chain
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR