Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPARD Rabbit Polyclonal Antibody

Supplier: Genetex
GTX11563
89286-012
Anti-PPARD Rabbit Polyclonal Antibody
WB: Use at a dilution of 1/1000. A starting antibody dilution of 1/5000 is recommended for chemiluminescent substrates. Higher antibody dilutions may be necessary for non-human samples. Detects, under reducing conditions, the zymogen form at 130-140 kDa, activated forms at 94-105 kDa (major bands), and breakdown products at 50 kDa, 34 kDa in cell culture media and lysates. EDTA/EGTA treatment of tissues or lysates is required to see latent zymogen. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PPARD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Pig, Rat

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty