Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FTCD Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MP6-XT22]

Supplier: Genetex
GTX11560
89317-302
Anti-FTCD Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MP6-XT22]
Recommended Starting Dilutions:Flow Cytometry: Neat - 1/10Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: Tnf
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation: FITC
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse, Rat

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty