Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL6R Rat Monoclonal Antibody [clone: JES6-1A12]

Supplier: Genetex
GTX11510
89317-300
Anti-IL6R Rat Monoclonal Antibody [clone: JES6-1A12]
Recommended Starting Dilutions:ELISA: 1 ug/ml - 4 ug/mlWestern Blotting: 1 ug/ml - 5 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL6R
Clonality: Monoclonal
Clone: JES6-1A12
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty