Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SORBS1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX11507
89316-564
Anti-SORBS1 Rabbit Polyclonal Antibody
IF: Use at a dilution of 1/500. WB: Use at a dilution of 1/2000. Detects a band of approximately 120 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SORBS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty