Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HAVCR2 Mouse Monoclonal Antibody [clone: 15F11]

Supplier: Genetex

GTX11506
89286-788EA 448.85 USD InStock
89286-788
Anti-HAVCR2 Mouse Monoclonal Antibody [clone: 15F11]
Antibodies
Type: Primary
Antigen: TIM3 / HAVCR2
Clonality: Monoclonal
Clone: 15F11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty