Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTCH1 Rat Monoclonal Antibody [clone: KT10b]

Supplier: Genetex
GTX11495
89315-936
Anti-PTCH1 Rat Monoclonal Antibody [clone: KT10b]
Type: Primary
Antigen: PTCH1
Clonality: Monoclonal
Clone: KT10b
Conjugation:
Epitope: Center
Host: Rat
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty