Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TMED10 Mouse Monoclonal Antibody [clone: BRIC 203]

Supplier: Genetex
GTX11463
89317-296
Anti-TMED10 Mouse Monoclonal Antibody [clone: BRIC 203]
Type: Primary
Antigen: TMED10
Clonality: Monoclonal
Clone: BRIC 203
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Dog, Rat

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty