Anti-PLIN1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX114627
89321-770EA 582.76 USD
89321-770
Anti-PLIN1 Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PLIN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR