Anti-ROBO3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-D9]

Supplier: GeneTex
GTX11457
89261-512EA 600.17 USD
89261-512
Anti-ROBO3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-D9]
Antibodies
Type: Primary
Antigen: ROBO3
Clonality: Monoclonal
Clone: B-D9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Center
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR