Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTF1A Mouse Monoclonal Antibody [clone: h.mcp.3]

Supplier: Genetex

GTX11453
89285-996EA 398.56 USD InStock
89285-996
Anti-PTF1A Mouse Monoclonal Antibody [clone: h.mcp.3]
Antibodies
Recommended Starting Dilutions:ELISA: Max Dilution 1/1000Functional Assays: Max Dilution 1/1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: PTF1A
Clonality: Monoclonal
Clone: h.mcp.3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Dog, Rat, Zebrafish

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty