Anti-NODAL Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11437
89317-292EA 640.44 USD
89317-292
Anti-NODAL Rabbit Polyclonal Antibody
Antibodies
Recommended Starting Dilutions:ICC/IF: 1 ug/mlWB: 1:500Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: NODAL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR