Anti-COL4A3BP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX114173
89321-504EA 582.76 USD
89321-504
Anti-COL4A3BP Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: 662-672 aa
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR