Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NOVA1 Mouse Monoclonal Antibody [clone: C10F5]

Supplier: Genetex
GTX11407
89317-288
Anti-NOVA1 Mouse Monoclonal Antibody [clone: C10F5]
Mouse monoclonal antibody [C10F5] to IFN alpha

Type: Primary
Antigen: NOVA1
Clonality: Monoclonal
Clone: C10F5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty