Anti-CEP290 / NPHP6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX114054
89321-460EA 582.76 USD
89321-460
Anti-CEP290 / NPHP6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Alcohol dehydrogenase 1B (alcohol dehydrogenase 1B (class I), beta polypeptide)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CEP290 / NPHP6
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR