Anti-GPR139 Mouse Monoclonal Antibody [clone: INN-hCG-68]

Supplier: GeneTex
GTX11391
89317-282EA 622.75 USD
89317-282
Anti-GPR139 Mouse Monoclonal Antibody [clone: INN-hCG-68]
Antibodies
Mouse monoclonal antibody [INN-hCG-68] to CG beta

Type: Primary
Antigen: CGB
Clonality: Monoclonal
Clone: INN-hCG-68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR