Anti-TFEB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113813
89321-398EA 582.76 USD
89321-398
Anti-TFEB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PSMD10 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 10)

  • Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1001. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TFEB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR