Anti-ABCE1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113748
89321-372EA 448.72 USD
89321-372
Anti-ABCE1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ICAM3(CD50) (intercellular adhesion molecule 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ABCE1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR