Anti-USP21 Rabbit Polyclonal Antibody

GTX113720
89321-366EA 376.76 USD
89321-366
Anti-USP21 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to ERp57 (protein disulfide isomerase family A, member 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: USP21
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR