Anti-APOA1BP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113627
89312-084EA 879.42 USD
89312-084
Anti-APOA1BP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to APOH (apolipoprotein H (beta-2-glycoprotein I))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: APOA1BP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR