Anti-CDH23 / USH1D Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113516
89321-298EA 468.44 USD
89321-298
Anti-CDH23 / USH1D Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Neuroserpin (serpin peptidase inhibitor, clade I (neuroserpin), member 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CDH23 / USH1D
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR