Anti-RGS13 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113351
89321-248EA 582.56 USD
89321-248
Anti-RGS13 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to STAT5A (signal transducer and activator of transcription 5A)

Suggested starting dilutions are as follows: WB: 1:500-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RGS13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR