Anti-ARG1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113215
89321-206EA 582.76 USD
89321-206
Anti-ARG1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ITK (IL2-inducible T-cell kinase)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ARG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR