Anti-EWS / EWSR1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113193
89321-192EA 582.76 USD
89321-192
Anti-EWS / EWSR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GATA1 (GATA binding protein 1 (globin transcription factor 1))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: EWS / EWSR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR