Anti-HIPPI Mouse Monoclonal Antibody [clone: 10651]

Supplier: GeneTex
GTX11311
89291-570EA 1196.84 USD
89291-570
Anti-HIPPI Mouse Monoclonal Antibody [clone: 10651]
Antibodies
Mouse monoclonal [2 - 28 - 33] to beta Tubulin

ICC: Use at an assay dependent dilution. IF: Use at a dilution of 1/1000, this dilution was determined by indirect immunofluorescent labeling of cultured human fibroblasts. WB: Use at an assay dependent dilution. Predicted molecular weight: 50 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HIPPI
Clonality: Monoclonal
Clone: 10651
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Bovine, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR