Anti-GPR81 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113043
89321-128EA 643.43 USD
89321-128
Anti-GPR81 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TRAIL (tumor necrosis factor (ligand) superfamily, member 10)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TNFSF10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR