Anti-PMAIP1 Rabbit Polyclonal Antibody

GTX113019
89321-122EA 376.43 USD
89321-122
Anti-PMAIP1 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to Thyroid Hormone Receptor alpha 2 (thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PMAIP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR