Anti-SLCO1B3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112684
89321-032EA 582.76 USD
89321-032
Anti-SLCO1B3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CD105 (endoglin)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SLCO1B3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR