Anti-USP16 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112593
89321-008EA 582.76 USD
89321-008
Anti-USP16 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IFIT3 (interferon-induced protein with tetratricopeptide repeats 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: USP16
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Horse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR