Anti-SEC23A Rabbit Polyclonal Antibody

GTX112401
89311-708EA 176.03 USD
89311-708
Anti-SEC23A Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to KIF1A (kinesin family member 1A)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SEC23A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Horse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR