Anti-PIAS2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112338
89320-952EA 582.76 USD
89320-952
Anti-PIAS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to beta Adducin (adducin 2 (beta))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PIAS2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: 185-196 aa
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR