Anti-FBXO4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112299
89320-944EA 582.76 USD
89320-944
Anti-FBXO4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to E2F8 (E2F transcription factor 8)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: E2F8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR