Anti-PON2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112191
89335-502EA 582.76 USD
89335-502
Anti-PON2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to G3BP1 (ras GTPase-activating protein-binding protein 1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PON2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR