Anti-LARGE Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112158
89320-918EA 582.76 USD
89320-918
Anti-LARGE Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to EPS8L2 (EPS8-like 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LARGE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: 421-433 aa
Host: Rabbit
Isotype:
Reactivity: Human, Bovine, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR