Anti-LAT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112026
89320-880EA 597.31 USD
89320-880
Anti-LAT Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to 14-3-3 theta (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Bovine, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR