Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-H3 Mouse Monoclonal Antibody [clone: VIN-11-5]

Supplier: Genetex

GTX11193
89291-566EA 665.23 USD InStock
89291-566
Anti-H3 Mouse Monoclonal Antibody [clone: VIN-11-5]
Antibodies
Mouse monoclonal [VIN - 11 - 5] to Vinculin

IF: Use at a dilution of 1/50. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: VCL
Clonality: Monoclonal
Clone: VIN-11-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty