Anti-GPR150 Rabbit Polyclonal Antibody

GTX111705
89311-448EA 176.03 USD
89311-448
Anti-GPR150 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Nuclear receptor subfamily 2 group E member 1 (nuclear receptor subfamily 2, group E, member 1)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: NR2E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR