Anti-HKR1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX111604
89320-770EA 582.76 USD
89320-770
Anti-HKR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PKM (muscle) (pyruvate kinase, muscle)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HKR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR