Anti-ARF4 Rabbit Polyclonal Antibody

GTX111593
89320-764EA 376.43 USD
89320-764
Anti-ARF4 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to PI 3 Kinase p85 beta (phosphoinositide-3-kinase, regulatory subunit 2 (beta))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ARF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR