Anti-NRIP3 Goat Polyclonal Antibody

Supplier: GeneTex
GTX11148
89317-880EA 705.93 USD
89317-880
Anti-NRIP3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Presenilin 2

Recommended Starting Dilutions: ELISA: Max Dilution 1/20,000 on immunising peptide. Immunohistology - Frozen: Max Dilution 1/1000. Immunohistology - Paraffin: Max Dilution 1/300. Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: NRIP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR