Anti-MED24 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX111445
89311-372EA 582.76 USD
89311-372
Anti-MED24 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TYK2 (tyrosine kinase 2)

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MED24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR