Anti-OR8H2 Rabbit Polyclonal Antibody

GTX111138
89320-644EA 376.43 USD
89320-644
Anti-OR8H2 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to TRAF2 (TNF receptor-associated factor 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR8H2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR