Anti-ATXN7L1 Rabbit Polyclonal Antibody

GTX111128
89311-260EA 176.03 USD
89311-260
Anti-ATXN7L1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to PPP2R2A (protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ATXN7L1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR