Anti-PPP1R3A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX111086
89320-628EA 582.76 USD
89320-628
Anti-PPP1R3A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PPIase (peptidylprolyl isomerase (cyclophilin)-like 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PPP1R3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR