Anti-IF3EI Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX111035
89320-612EA 582.76 USD
89320-612
Anti-IF3EI Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ZNF346 (zinc finger protein 346)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: IF3EI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR