Anti-MRPL54 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX110976
89311-208EA 583.32 USD
89311-208
Anti-MRPL54 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RPRM (reprimo, TP53 dependent G2 arrest mediator candidate)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: MRPL54
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR