Anti-ARSD Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX110775
89311-186EA 582.76 USD
89311-186
Anti-ARSD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CBLC (Cas-Br-M (murine) ecotropic retroviral transforming sequence c)

  • Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ARSD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR