Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IRX2 Mouse Monoclonal Antibody [clone: INN-hFSH-60]

Supplier: Genetex

GTX11069
89285-988
Anti-IRX2 Mouse Monoclonal Antibody [clone: INN-hFSH-60]
Mouse monoclonal antibody [INN-hFSH-60] to FSH beta

Type: Primary
Antigen: IRX2
Clonality: Monoclonal
Clone: INN-hFSH-60
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty