Anti-OR5H6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX110684
89320-572EA 582.76 USD
89320-572
Anti-OR5H6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to VCAM1 (vascular cell adhesion molecule 1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR5H6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR