Anti-OPN4 Rabbit Polyclonal Antibody

GTX110661
89334-218EA 180.24 USD
89334-218
Anti-OPN4 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to HSBP1 (heat shock factor binding protein 1)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: OPN4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR